Assay Details
| Gene Symbol: Arf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007478.3 | NP_031504.1 | 3 - 4 | 489 | 68 | |
| GenBank mRNA | AK151296.1 | - | 3 - 4 | 583 | 68 | |
| AK154948.1 | - | 3 - 4 | 537 | 68 | ||
| AK165176.1 | - | 3 - 4 | 543 | 68 | ||
| BC014778.1 | - | 3 - 4 | 489 | 3582243 | 68 | |
| BC024935.1 | - | 3 - 4 | 534 | 3969152 | 68 | |
| D87900.1 | - | 3 - 4 | 458 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arf3
Genomic Map