Assay Details
| Gene Symbol: Hsd17b4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008292.4 | NP_032318.2 | 2 - 3 | 257 | 64 | |
| GenBank mRNA | AK004866.1 | - | 2 - 3 | 257 | 64 | |
| AK088381.1 | - | 2 - 3 | 232 | 64 | ||
| AK147182.1 | - | 2 - 3 | 229 | 64 | ||
| AK152532.1 | - | 2 - 3 | 152 | 64 | ||
| AK161655.1 | - | 2 - 3 | 152 | 64 | ||
| AK166801.1 | - | 2 - 3 | 170 | 64 | ||
| AK167060.1 | - | 2 - 3 | 207 | 64 | ||
| AK169077.1 | - | 2 - 3 | 159 | 64 | ||
| AK205578.1 | - | 1 - 2 | 56 | 64 | ||
| X89998.1 | - | 2 - 3 | 118 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hsd17b4
Genomic Map