Assay Details
| Gene Symbol: Hsd17b4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008292.4 | NP_032318.2 | 8 - 9 | 773 | 56 | |
| GenBank mRNA | AK004866.1 | - | 8 - 9 | 773 | 56 | |
| AK088381.1 | - | 8 - 9 | 748 | 56 | ||
| AK147182.1 | - | 8 - 9 | 745 | 56 | ||
| AK152532.1 | - | 8 - 9 | 668 | 56 | ||
| AK161655.1 | - | 8 - 9 | 668 | 56 | ||
| AK166801.1 | - | 8 - 9 | 686 | 56 | ||
| AK167060.1 | - | 8 - 9 | 723 | 56 | ||
| AK169077.1 | - | 8 - 9 | 675 | 56 | ||
| BC022175.1 | - | 8 - 9 | 646 | 4209394 | 56 | |
| X89998.1 | - | 8 - 9 | 634 | 56 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hsd17b4
Genomic Map