Assay Details
Gene Symbol: Slc3a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161413.1 | NP_001154885.1 | 2 - 3 | 794 | 78 | |
NM_008577.4 | NP_032603.3 | 1 - 2 | 553 | 78 | ||
GenBank mRNA | AB023408.1 | - | 1 - 2 | 508 | 78 | |
AK090046.1 | - | 1 - 2 | 531 | 78 | ||
AK161280.1 | - | 2 - 3 | 795 | 78 | ||
AK165417.1 | - | 1 - 2 | 535 | 78 | ||
AK165550.1 | - | 1 - 2 | 531 | 78 | ||
BC065173.1 | - | 1 - 2 | 513 | 6415269 | 78 | |
BU054912.1 | - | 1 - 2 | 555 | 78 | ||
U25708.1 | - | 1 - 2 | 530 | 78 | ||
X14309.1 | - | 1 - 2 | 495 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
