Assay Details
Gene Symbol: Slc3a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161413.1 | NP_001154885.1 | 8 - 9 | 1516 | 87 | |
NM_008577.4 | NP_032603.3 | 7 - 8 | 1275 | 87 | ||
GenBank mRNA | AB023408.1 | - | 7 - 8 | 1230 | 87 | |
AK090046.1 | - | 7 - 8 | 1253 | 87 | ||
AK156285.1 | - | 7 - 8 | 747 | 87 | ||
AK161280.1 | - | 8 - 9 | 1517 | 87 | ||
AK165417.1 | - | 7 - 8 | 1257 | 87 | ||
AK165550.1 | - | 7 - 8 | 1253 | 87 | ||
AK197569.1 | - | 1 - 2 | 107 | 87 | ||
AK206213.1 | - | 2 - 3 | 216 | 87 | ||
AK210586.1 | - | 1 - 2 | 107 | 87 | ||
BC065173.1 | - | 7 - 8 | 1235 | 6415269 | 87 | |
X14309.1 | - | 7 - 8 | 1217 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
