Assay Details
Gene Symbol: Map7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198635.1 | NP_001185564.1 | 1 - 2 | 159 | 65 | |
NM_008635.2 | NP_032661.2 | 1 - 2 | 159 | 65 | ||
GenBank mRNA | AB098611.1 | - | 1 - 2 | 65 | 65 | |
AK014575.1 | - | 1 - 2 | 77 | 65 | ||
BC042771.1 | - | 1 - 2 | 84 | 65 | ||
BC044733.1 | - | 1 - 2 | 84 | 65 | ||
BC046555.1 | - | 1 - 2 | 84 | 65 | ||
BC052637.1 | - | 1 - 2 | 95 | 6467153 | 65 | |
CJ074623.1 | - | 1 - 2 | 136 | 65 | ||
Y15197.1 | - | 1 - 2 | 65 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
