Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400031656.mm.1 |
TC0400003841.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400031656.mm.1 |
NM_008675.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400031656.mm.1 |
NM_008675.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400031655.mm.1 |
TC0400003841.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400031655.mm.1 |
NM_008675.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400031655.mm.1 |
NM_008675.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400031662.mm.1 |
TC0400003841.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400031662.mm.1 |
NM_008675.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400031662.mm.1 |
NM_008675.2 |
Refseq |