Assay Details
Gene Symbol: Nfkbie | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF030896.1 | - | 4 - 5 | 782 | 77 | |
AK008843.1 | - | 4 - 5 | 1021 | 77 | ||
AK011965.1 | - | 4 - 5 | 1013 | 77 | ||
AK088585.1 | - | 3 - 4 | 1595 | 77 | ||
AK151450.1 | - | 4 - 5 | 1021 | 77 | ||
AK151692.1 | - | 4 - 5 | 1021 | 77 | ||
AK153248.1 | - | 4 - 5 | 988 | 77 | ||
BC030923.1 | - | 4 - 5 | 1007 | 4909950 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
