Assay Details
Gene Symbol: Nmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008707.3 | NP_032733.1 | 4 - 5 | 514 | 65 | |
GenBank mRNA | AF043326.1 | - | 4 - 5 | 526 | 65 | |
AK146523.1 | - | 4 - 5 | 529 | 65 | ||
AK213723.1 | - | 2 - 3 | 155 | 65 | ||
BC016526.1 | - | 4 - 5 | 514 | 3589678 | 65 | |
BC021635.1 | - | 4 - 5 | 528 | 3989540 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:

Citations (1)
N-myristoyltransferase 1 is essential in early mouse development.
- Authors
- Yang SH, Shrivastav A, Kosinski C, Sharma RK, Chen MH, Berthiaume LG, Peters LL, Chuang PT, Young SG, Bergo MO
- Journal
- J Biol Chem - Page No 18990 (Fri May 13 00:00:00 EDT 2005)
- Volume (Year)
- 280 (2005)
- Product Usage
- -
