Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300017225.mm.1 |
NM_008756.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300017225.mm.1 |
NM_008756.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300017225.mm.1 |
TC1300002536.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300017229.mm.1 |
NM_008756.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300017229.mm.1 |
NM_008756.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300017229.mm.1 |
TC1300002536.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300017226.mm.1 |
NM_008756.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300017226.mm.1 |
NM_008756.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300017226.mm.1 |
TC1300002536.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300017228.mm.1 |
NM_008756.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300017228.mm.1 |
NM_008756.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300017228.mm.1 |
TC1300002536.mm.1 |
MTA TC |