Assay Details
Gene Symbol: Cntn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008779.2 | NP_032805.2 | 5 - 6 | 740 | 92 | |
GenBank mRNA | L01991.1 | - | 849 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
