Assay Details
| Gene Symbol: Cntn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008779.2 | NP_032805.2 | 6 - 7 | 846 | 89 | |
| GenBank mRNA | L01991.1 | - | 955 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cntn3
Genomic Map