Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500005029.mm.1 |
NM_001045558.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500005029.mm.1 |
NM_001045558.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500005029.mm.1 |
NM_008837.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500005029.mm.1 |
TC1500000670.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500005032.mm.1 |
NM_001045558.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500005032.mm.1 |
NM_001045558.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500005032.mm.1 |
NM_008837.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500005032.mm.1 |
TC1500000670.mm.1 |
MTA TC |