Assay Details
Gene Symbol: Ptpn22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008979.2 | NP_033005.1 | 12 - 13 | 1124 | 66 | |
NR_104070.1 | 14 - 15 | 1149 | 66 | |||
GenBank mRNA | AK153812.1 | - | 14 - 15 | 1150 | 66 | |
BC055377.1 | - | 12 - 13 | 1046 | 30077916 | 66 | |
M90388.1 | - | 12 - 13 | 1109 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
