Assay Details
| Gene Symbol: Aldh1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009022.4 | NP_033048.2 | 8 - 9 | 961 | 101 | |
| GenBank mRNA | AK078553.1 | - | 8 - 9 | 902 | 101 | |
| BC075704.1 | - | 8 - 9 | 928 | 30471325 | 101 | |
| X99273.1 | - | 8 - 9 | 906 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aldh1a2
Genomic Map