Assay Details
| Gene Symbol: Rgs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009061.4 | NP_033087.2 | 2 - 3 | 247 | 87 | |
| GenBank mRNA | AF215668.1 | - | 2 - 3 | 232 | 87 | |
| AF432916.1 | - | 2 - 3 | 226 | 87 | ||
| AK029310.1 | - | 2 - 3 | 249 | 87 | ||
| AK031603.1 | - | 2 - 3 | 249 | 87 | ||
| AK077922.1 | - | 2 - 3 | 248 | 87 | ||
| AK162276.1 | - | 2 - 3 | 249 | 87 | ||
| BC023001.1 | - | 2 - 3 | 230 | 5365271 | 87 | |
| U67187.1 | - | 2 - 3 | 236 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rgs2
Genomic Map