Assay Details
| Gene Symbol: S100a10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK170919.1 | - | 2 - 3 | 228 | 77 | |
| BC025044.1 | - | 2 - 3 | 179 | 4949465 | 77 | |
| M16465.1 | - | 2 - 3 | 201 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

S100a10
Genomic Map