Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500003149.mm.1 |
TC0500000344.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500003149.mm.1 |
NM_001205132.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500003149.mm.1 |
NM_001205132.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500003149.mm.1 |
NM_001205133.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500003149.mm.1 |
NM_001205133.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500003149.mm.1 |
NM_009535.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500003149.mm.1 |
NM_009535.3 |
Refseq |