Assay Details
Gene Symbol: Coq7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF053770.1 | - | 3 - 4 | 450 | 67 | |
AF080580.1 | - | 4 - 5 | 513 | 67 | ||
AF098949.1 | - | 4 - 5 | 579 | 67 | ||
AK158380.1 | - | 4 - 5 | 564 | 67 | ||
BC028276.1 | - | 2 - 3 | 255 | 67 | ||
BC038681.1 | - | 4 - 5 | 546 | 1380996 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25458767 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25458765 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25458764 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25458766 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25458769 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25458768 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P161691 | AF098949.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P161691 | BC028276.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P161691 | NM_009940.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P161691 | BC038681.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P161691 | AF098949.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P161691 | BC028276.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P161691 | NM_009940.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P161691 | BC038681.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P137134 | AK158380.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 670312 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 624350 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 36877 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 373468 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 426456 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 428706 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 40144 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P122620 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P119224 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P137134 | AK158380.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P161691 | AF098949.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P161691 | BC028276.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P161691 | NM_009940.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P161691 | BC038681.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P161691 | AF098949.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P161691 | BC028276.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P161691 | NM_009940.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P161691 | BC038681.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25458777 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25458770 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25458779 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25458775 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25458773 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25458757 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25458754 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25458780 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25458755 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25458759 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25458768 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25458764 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25458771 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25458766 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25458761 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25458782 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25458758 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25458772 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25458762 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P119224 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01616384 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06339878 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P122620 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01616392 | - | - |
Whole Mouse Genome G4122A | A_51_P161691 | AF098949.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P161691 | BC028276.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P161691 | NM_009940.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P161691 | BC038681.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P161691 | AF098949.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P161691 | BC028276.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P161691 | NM_009940.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P161691 | BC038681.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P161682 | U81277.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P161682 | AF053770.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P161682 | AF080580.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P161682 | AF098949.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P161682 | AK158380.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128833 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P122620 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P119224 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06367992 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06367990 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06367996 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06368001 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06367994 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25458768 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25458767 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25458766 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25458769 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25458765 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25458764 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458700 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458687 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458734 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458742 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458757 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458728 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458686 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458744 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458737 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458659 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458676 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458721 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458739 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458736 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458771 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458777 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458746 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458766 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458720 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458782 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458775 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458674 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458779 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458761 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458781 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458673 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458731 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458748 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458712 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458689 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458664 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458698 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458768 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458725 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458745 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458770 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458684 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458708 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458682 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458696 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458677 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458691 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458740 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458774 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458717 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458754 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458741 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458665 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458719 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458732 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458706 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458758 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458662 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458772 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458723 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458680 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458751 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458716 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458764 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458714 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458697 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458703 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25458709 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25458751 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25458738 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25458758 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25458737 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25458610 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25458774 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25458759 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25458764 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25458744 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25458761 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25458608 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25458770 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25458782 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25458604 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25458736 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06353231 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25458734 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25458742 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25458682 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25458632 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06437242 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06479177 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06507552 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06363295 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25458732 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25458687 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06115008 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25458613 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06479182 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25458777 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06082133 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06479183 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25458775 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25458780 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25458772 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25458607 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25458748 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25458691 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25458731 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06385822 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25458773 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25458612 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25458684 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25458625 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25458689 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25458624 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25458728 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25458601 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25458686 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25458739 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25458611 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25458768 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06253547 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06507551 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25458721 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25458693 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25458630 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25458679 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25458771 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06363298 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06645555 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25458743 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06392058 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25458781 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06067538 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25458628 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10567307 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10567309 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10567311 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10567310 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10567308 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10567312 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10567314 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10567313 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10567315 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10567306 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1416665_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10567303 | - | - |
Mouse Genome 430 2.0 Array | 1416665_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1416665_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12850_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12850_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0700034685.mm.1 | NM_009940.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700034685.mm.1 | TC0700004173.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700034687.mm.1 | NM_009940.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700034687.mm.1 | TC0700004173.mm.1 | MTA TC |
Mouse Expression Set 430 | 1416665_PM_at | - | - |
Mouse Expression Set 430 | 1416665_at | - | - |
Murine Genome U74v2 Set | 93582_at | - | - |
Murine Genome U74v2 Set | 93582_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1416665_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1416665_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1416665_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1416665_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 631704 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 3190632 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003190632 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2729263 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2729263 | - | - |
mouse-6 v1.1 (Illumina) | scl30756.4.28_1-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3190632 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl30756.4.28_1-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3190632 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2729263 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2729263 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl30756.4.28_1-S | - | - |
