Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300020561.mm.1 |
NM_010432.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020561.mm.1 |
NM_010432.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020561.mm.1 |
TC0300002630.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300020563.mm.1 |
NM_010432.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020563.mm.1 |
NM_010432.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020563.mm.1 |
TC0300002630.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300020566.mm.1 |
NM_010432.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020566.mm.1 |
NM_010432.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020566.mm.1 |
TC0300002630.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300020567.mm.1 |
NM_010432.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020567.mm.1 |
NM_010432.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020567.mm.1 |
TC0300002630.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300020562.mm.1 |
NM_010432.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020562.mm.1 |
NM_010432.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020562.mm.1 |
TC0300002630.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300020565.mm.1 |
NM_010432.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020565.mm.1 |
NM_010432.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020565.mm.1 |
TC0300002630.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300020564.mm.1 |
NM_010432.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020564.mm.1 |
NM_010432.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020564.mm.1 |
TC0300002630.mm.1 |
MTA TC |