Assay Details
| Gene Symbol: Ogg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010957.4 | NP_035087.3 | 4 - 5 | 999 | 90 | |
| GenBank mRNA | AF000669.1 | - | 4 - 5 | 877 | 90 | |
| AK146870.1 | - | 4 - 5 | 999 | 90 | ||
| AK165875.1 | - | 4 - 5 | 951 | 90 | ||
| AK167456.1 | - | 4 - 5 | 1059 | 90 | ||
| BC138733.1 | - | 4 - 5 | 955 | 8861755 | 90 | |
| BC138736.1 | - | 4 - 5 | 955 | 8861758 | 90 | |
| U96711.1 | - | 4 - 5 | 964 | 90 | ||
| Y11247.1 | - | 4 - 5 | 930 | 90 | ||
| Y13479.1 | - | 4 - 5 | 1000 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ogg1
Genomic Map