Assay Details
Gene Symbol: Nt5e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011851.4 | NP_035981.1 | 6 - 7 | 1294 | 96 | |
GenBank mRNA | AK028723.1 | - | 6 - 7 | 1264 | 96 | |
AK029979.1 | - | 6 - 7 | 1264 | 96 | ||
AK128997.1 | - | 6 - 7 | 1263 | 96 | ||
AK154614.1 | - | 6 - 7 | 1271 | 96 | ||
BC119268.1 | - | 6 - 7 | 1238 | 8734017 | 96 | |
BC119270.1 | - | 6 - 7 | 1238 | 8734019 | 96 | |
L12059.1 | - | 6 - 7 | 1217 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
