Assay Details
| Gene Symbol: Pde6g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF190928.1 | - | 1 - 2 | 150 | 101 | |
| AK080735.1 | - | 2 - 3 | 296 | 101 | ||
| BC082288.1 | - | 2 - 3 | 253 | 6490841 | 101 | |
| Y00746.1 | - | 2 - 3 | 271 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pde6g
Genomic Map