Assay Details
| Gene Symbol: Gnb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013531.4 | NP_038559.2 | 7 - 8 | 638 | 86 | |
| GenBank mRNA | AF277893.1 | - | 6 - 7 | 502 | 86 | |
| AF501886.1 | - | 6 - 7 | 502 | 86 | ||
| AK036816.1 | - | 7 - 8 | 640 | 86 | ||
| AK098139.1 | - | 7 - 8 | 571 | 86 | ||
| AK152227.1 | - | 7 - 8 | 629 | 86 | ||
| AK155309.1 | - | 7 - 8 | 697 | 86 | ||
| AK158965.1 | - | 8 - 9 | 868 | 86 | ||
| AK167416.1 | - | 7 - 8 | 671 | 86 | ||
| AK168302.1 | - | 7 - 8 | 594 | 86 | ||
| AK172147.1 | - | 8 - 9 | 771 | 86 | ||
| AK195471.1 | - | 2 - 3 | 171 | 87 | ||
| AK211063.1 | - | 2 - 3 | 171 | 87 | ||
| BC028753.1 | - | 7 - 8 | 609 | 1348753 | 86 | |
| M63658.1 | - | 6 - 7 | 542 | 86 | ||
| M87286.1 | - | 6 - 7 | 542 | 86 | ||
| S86124.1 | - | 6 - 7 | 542 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gnb4
Genomic Map