Assay Details
| Gene Symbol: Cpxm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018867.5 | NP_061355.3 | 4 - 5 | 802 | 102 | |
| GenBank mRNA | AF017639.1 | - | 4 - 5 | 803 | 102 | |
| AK019509.1 | - | 4 - 5 | 801 | 102 | ||
| AK038874.1 | - | 4 - 5 | 685 | 102 | ||
| BC021444.1 | - | 4 - 5 | 724 | 5038528 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cpxm2
Genomic Map