Assay Details
| Gene Symbol: Sae1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285891.1 | NP_001272820.1 | 3 - 4 | 540 | 59 | |
| NM_001285892.1 | NP_001272821.1 | 3 - 4 | 540 | 59 | ||
| NM_019748.3 | NP_062722.1 | 3 - 4 | 540 | 59 | ||
| GenBank mRNA | AB024303.1 | - | 3 - 4 | 428 | 59 | |
| AK010313.1 | - | 3 - 4 | 417 | 59 | ||
| AK011772.1 | - | 3 - 4 | 451 | 59 | ||
| AK087556.1 | - | 3 - 4 | 431 | 59 | ||
| AK090012.1 | - | 3 - 4 | 59 | |||
| AK154139.1 | - | 3 - 4 | 453 | 59 | ||
| AK159672.1 | - | 3 - 4 | 442 | 59 | ||
| AK162789.1 | - | 3 - 4 | 428 | 59 | ||
| BC068164.1 | - | 3 - 4 | 429 | 30359189 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sae1
Genomic Map