Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200014513.mm.1 |
NM_001162503.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014513.mm.1 |
NM_001162503.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014513.mm.1 |
NM_021507.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014513.mm.1 |
NM_021507.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014513.mm.1 |
NR_027888.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014513.mm.1 |
NR_027888.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014513.mm.1 |
TC0200001859.mm.1 |
MTA TC |