Assay Details
Gene Symbol: Clstn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022319.2 | NP_071714.2 | 5 - 6 | 987 | 68 | |
GenBank mRNA | AJ278069.1 | - | 5 - 6 | 935 | 68 | |
AK220500.1 | - | 5 - 6 | 1001 | 68 | ||
BC063058.1 | - | 5 - 6 | 987 | 5705591 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
