Assay Details
| Gene Symbol: Igf2bp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023670.3 | NP_076159.3 | 5 - 6 | 881 | 114 | |
| GenBank mRNA | AB046173.1 | - | 5 - 6 | 398 | 114 | |
| AK011689.1 | - | 5 - 6 | 662 | 114 | ||
| AK030096.1 | - | 5 - 6 | 741 | 114 | ||
| AK088465.1 | - | 5 - 6 | 640 | 114 | ||
| AK147140.1 | - | 5 - 6 | 885 | 114 | ||
| BC045138.1 | - | 5 - 6 | 511 | 6306448 | 114 | |
| BC049082.1 | - | 5 - 6 | 547 | 5706238 | 114 | |
More Information
Updated Mapping Information
Set Membership: |
|

Igf2bp3
Genomic Map