Assay Details
| Gene Symbol: Igf2bp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023670.3 | NP_076159.3 | 8 - 9 | 1423 | 62 | |
| GenBank mRNA | AB046173.1 | - | 8 - 9 | 940 | 62 | |
| AK011689.1 | - | 8 - 9 | 1204 | 62 | ||
| AK030096.1 | - | 7 - 8 | 2744 | 62 | ||
| AK088465.1 | - | 8 - 9 | 1182 | 62 | ||
| AK147140.1 | - | 8 - 9 | 1427 | 62 | ||
| BC045138.1 | - | 8 - 9 | 1053 | 6306448 | 62 | |
| BC049082.1 | - | 8 - 9 | 1089 | 5706238 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Igf2bp3
Genomic Map