Assay Details
Gene Symbol: Atp6v0e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025272.2 | NP_079548.1 | 1 - 2 | 177 | 76 | |
GenBank mRNA | AK007610.1 | - | 1 - 2 | 156 | 76 | |
AK008464.1 | - | 1 - 2 | 157 | 76 | ||
AK013229.1 | - | 1 - 2 | 138 | 76 | ||
AK017760.1 | - | 1 - 2 | 179 | 76 | ||
AK030335.1 | - | 1 - 2 | 129 | 76 | ||
AK134836.1 | - | 1 - 2 | 161 | 76 | ||
AK159669.1 | - | 1 - 2 | 165 | 76 | ||
AK168015.1 | - | 1 - 2 | 146 | 76 | ||
BC024733.1 | - | 1 - 2 | 141 | 5374017 | 76 | |
BY707658.1 | - | 1 - 2 | 153 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
