| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100036646.mm.1 |
NM_025310.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100036646.mm.1 |
NM_025310.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100036646.mm.1 |
TC1100003917.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100036647.mm.1 |
NM_025310.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100036647.mm.1 |
NM_025310.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100036647.mm.1 |
TC1100003917.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100036645.mm.1 |
NM_025310.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100036645.mm.1 |
NM_025310.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100036645.mm.1 |
TC1100003917.mm.1 |
MTA TC |