Assay Details
Gene Symbol: Hsd17b14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025330.3 | NP_079606.3 | 7 - 8 | 593 | 92 | |
GenBank mRNA | AK002831.1 | - | 7 - 8 | 546 | 92 | |
AK010556.1 | - | 3 - 4 | 258 | 92 | ||
BC064042.1 | - | 3 - 4 | 376 | 1332242 | 92 | |
BC099530.1 | - | 3 - 4 | 262 | 92 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK010556.1, BC064042.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
