Assay Details
| Gene Symbol: Pgd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081274.1 | NP_001074743.1 | 11 - 12 | 1321 | 110 | |
| GenBank mRNA | AK002894.1 | - | 11 - 12 | 1283 | 110 | |
| AK004217.1 | - | 4 - 5 | 525 | 110 | ||
| AK077110.1 | - | 11 - 12 | 1280 | 110 | ||
| AK145602.1 | - | 11 - 12 | 1285 | 110 | ||
| AK150210.1 | - | 11 - 12 | 1284 | 110 | ||
| AK150490.1 | - | 11 - 12 | 1280 | 110 | ||
| AK151380.1 | - | 11 - 12 | 1280 | 110 | ||
| AK151993.1 | - | 11 - 12 | 1280 | 110 | ||
| AK152758.1 | - | 11 - 12 | 1280 | 110 | ||
| AK152894.1 | - | 11 - 12 | 1280 | 110 | ||
| AK153409.1 | - | 11 - 12 | 1277 | 110 | ||
| AK155027.1 | - | 11 - 12 | 1280 | 110 | ||
| AK166733.1 | - | 11 - 12 | 1275 | 110 | ||
| AK166947.1 | - | 11 - 12 | 1257 | 110 | ||
| AK167215.1 | - | 11 - 12 | 1275 | 110 | ||
| AK168251.1 | - | 11 - 12 | 1280 | 110 | ||
| BC008646.1 | - | 11 - 12 | 1252 | 3155147 | 110 | |
| BC011329.1 | - | 11 - 12 | 1270 | 3984654 | 110 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pgd
Genomic Map