Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700006850.mm.1 |
TC0700000819.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700006850.mm.1 |
NM_001253920.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006850.mm.1 |
NM_001253920.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006850.mm.1 |
NM_026270.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006850.mm.1 |
NM_026270.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006835.mm.1 |
TC0700000819.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700006835.mm.1 |
NM_026270.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006835.mm.1 |
NM_026270.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006849.mm.1 |
TC0700000819.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700006849.mm.1 |
NM_001253920.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006849.mm.1 |
NM_001253920.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006849.mm.1 |
NM_026270.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006849.mm.1 |
NM_026270.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006836.mm.1 |
TC0700000819.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700006836.mm.1 |
NM_026270.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006836.mm.1 |
NM_026270.4 |
Refseq |