Assay Details
| Gene Symbol: Nudt12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026497.2 | NP_080773.1 | 2 - 3 | 318 | 64 | |
| GenBank mRNA | AK002641.1 | - | 2 - 3 | 319 | 64 | |
| AK083846.1 | - | 2 - 3 | 276 | 64 | ||
| AK147867.1 | - | 2 - 3 | 272 | 64 | ||
More Information
Set Membership: |
|

Nudt12
Genomic Map