Assay Details
| Gene Symbol: Ict1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026729.1 | NP_081005.1 | 3 - 4 | 211 | 87 | |
| GenBank mRNA | AK003192.1 | - | 3 - 4 | 211 | 87 | |
| AK003286.1 | - | 3 - 4 | 183 | 87 | ||
| BC028523.1 | - | 3 - 4 | 312 | 2938122 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ict1
Genomic Map