Assay Details
Gene Symbol: Acy3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF356878.1 | - | 1 - 2 | 42 | 57 | |
AK002247.1 | - | 1 - 2 | 41 | 57 | ||
AK143782.1 | - | 1 - 2 | 44 | 57 | ||
AY169234.1 | - | 1 - 2 | 41 | 57 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF356878.1, AK002247.1, AK143782.1, AY169234.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
