Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100040631.mm.1 |
NM_027874.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100040631.mm.1 |
NM_027874.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100040631.mm.1 |
NM_139059.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100040631.mm.1 |
NM_139059.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100040631.mm.1 |
TC1100004236.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100040633.mm.1 |
NM_027874.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100040633.mm.1 |
NM_027874.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100040633.mm.1 |
NM_139059.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100040633.mm.1 |
NM_139059.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100040633.mm.1 |
TC1100004236.mm.1 |
MTA TC |