Assay Details
Gene Symbol: Csnk1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027874.2 | NP_082150.1 | 5 - 6 | 1060 | 66 | |
NM_139059.2 | NP_620690.1 | 5 - 6 | 1060 | 66 | ||
GenBank mRNA | AK004606.1 | - | 5 - 6 | 820 | 66 | |
AK051052.1 | - | 5 - 6 | 1059 | 66 | ||
AK088642.1 | - | 5 - 6 | 795 | 66 | ||
AK140317.1 | - | 5 - 6 | 785 | 66 | ||
AK143534.1 | - | 5 - 6 | 761 | 66 | ||
AK146600.1 | - | 5 - 6 | 1050 | 66 | ||
AK150387.1 | - | 5 - 6 | 789 | 66 | ||
AK150815.1 | - | 5 - 6 | 787 | 66 | ||
AK152721.1 | - | 5 - 6 | 1002 | 66 | ||
AK157812.1 | - | 5 - 6 | 795 | 66 | ||
AK180857.1 | - | 1 - 2 | 135 | 66 | ||
AK218352.1 | - | 1 - 2 | 84 | 66 | ||
BC004604.1 | - | 5 - 6 | 1046 | 3585286 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
