Assay Details
| Gene Symbol: Telo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163661.1 | NP_001157133.1 | 16 - 17 | 2603 | 90 | |
| NM_027880.2 | NP_082156.2 | 16 - 17 | 2514 | 90 | ||
| GenBank mRNA | AK004582.1 | - | 16 - 17 | 2384 | 90 | |
| AK028658.1 | - | 16 - 17 | 2368 | 90 | ||
| AK050773.1 | - | 16 - 17 | 2333 | 90 | ||
| AK143827.1 | - | 16 - 17 | 2604 | 90 | ||
| AK146963.1 | - | 16 - 17 | 2390 | 90 | ||
| AK154727.1 | - | 16 - 17 | 2416 | 90 | ||
| AK158181.1 | - | 5 - 6 | 90 | |||
More Information
Set Membership: |
|

Telo2
Genomic Map