Assay Details
| Gene Symbol: Ppp1r14d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290796.1 | NP_001277725.1 | 4 - 5 | 505 | 89 | |
| NM_028104.4 | NP_082380.1 | 3 - 4 | 440 | 89 | ||
| GenBank mRNA | AF408400.2 | - | 3 - 4 | 439 | 89 | |
| AK008348.1 | - | 3 - 4 | 398 | 89 | ||
| AY179331.1 | - | 4 - 5 | 492 | 89 | ||
| BC024547.1 | - | 3 - 4 | 439 | 4989982 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21568531 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21568533 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21568534 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2009673 | NM_028104.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2009673 | AK008348.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2009673 | BC024547.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 576452 | AK039069.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 576452 | AK039075.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 510094 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 40579 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 546305 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 851221 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 820574 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 35473 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 446763 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 43250 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 173320 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P121243 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P139472 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2009673 | NM_028104.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2009673 | AK008348.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2009673 | BC024547.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568481 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568518 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568501 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568497 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568514 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568527 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568525 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568508 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568483 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568516 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568531 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568491 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568513 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568522 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568500 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568507 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568510 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568515 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568529 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568480 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568489 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568495 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568486 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568498 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568496 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568534 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568485 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568493 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568520 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568503 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568484 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568505 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568494 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04664589 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04664598 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P139472 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P121243 | - | - |
| Whole Mouse Genome G4122A | A_52_P40345 | NM_028104.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P40345 | AF408400.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P40345 | AK008348.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P40345 | AY179331.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P139472 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P121243 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P04664589 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01350840 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02259050 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02259046 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01350824 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02259054 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02259047 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01350842 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02259052 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01350825 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21568531 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21568533 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21568534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568412 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01747716 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568494 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02259046 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568510 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568475 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01566903 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02180012 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02259053 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01970355 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568473 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01429611 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01151628 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568511 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568480 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568465 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01479279 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568495 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01328088 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568490 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568462 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568500 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568471 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568470 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568507 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568481 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568485 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568469 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568484 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568472 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568498 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568464 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568476 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568466 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10486195 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486192 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486193 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486196 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486194 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417990_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10486191 | - | - |
| Mouse Genome 430 2.0 Array | 1417990_at | - | - |
| Mouse Genome 430 2.0 Array | 1417990_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72112_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72112_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200036978.mm.1 | NM_028104.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200036978.mm.1 | TC0200004425.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200036977.mm.1 | NM_028104.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200036977.mm.1 | TC0200004425.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417990_PM_at | - | - |
| Mouse Expression Set 430 | 1417990_at | - | - |
| Murine Genome U74v2 Set | 165491_at | - | - |
| Murine Genome U74v2 Set | 165491_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417990_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417990_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417990_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417990_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 911946 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2190168 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002190168 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1253015 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1253015 | - | - |
| mouse-6 v1.1 (Illumina) | scl18808.6.1_35-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2190168 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl18808.6.1_35-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2190168 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2872371 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1253015 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2872371 | - | - |

Ppp1r14d
Genomic Map