Assay Details
Gene Symbol: Ift57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028680.3 | NP_082956.2 | 4 - 5 | 632 | 106 | |
GenBank mRNA | AF274590.1 | - | 4 - 5 | 650 | 106 | |
AK047217.1 | - | 4 - 5 | 632 | 106 | ||
BC138143.1 | - | 4 - 5 | 606 | 8861162 | 106 | |
BC138144.1 | - | 4 - 5 | 606 | 8861163 | 106 | |
DQ266091.1 | - | 4 - 5 | 673 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
