Assay Details
| Gene Symbol: Ift57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028680.3 | NP_082956.2 | 7 - 8 | 889 | 95 | |
| GenBank mRNA | AF274590.1 | - | 7 - 8 | 907 | 95 | |
| AK014731.1 | - | 7 - 8 | 941 | 95 | ||
| AK040328.1 | - | 4 - 5 | 460 | 95 | ||
| AK047217.1 | - | 7 - 8 | 889 | 95 | ||
| BC138143.1 | - | 7 - 8 | 863 | 8861162 | 95 | |
| BC138144.1 | - | 7 - 8 | 863 | 8861163 | 95 | |
| DQ266091.1 | - | 7 - 8 | 930 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ift57
Genomic Map