Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000010444.mm.1 |
NM_001277292.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010444.mm.1 |
NM_001277293.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010444.mm.1 |
NM_001277294.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010444.mm.1 |
NM_001277295.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010444.mm.1 |
NM_028736.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010444.mm.1 |
NM_130891.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010444.mm.1 |
NM_133442.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010444.mm.1 |
NR_102387.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010444.mm.1 |
TC1000001466.mm.1 |
MTA TC |