| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1500014855.mm.1 |
NM_028763.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014855.mm.1 |
NM_028763.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014855.mm.1 |
TC1500001887.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500014861.mm.1 |
NM_001013362.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014861.mm.1 |
NM_001013362.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014861.mm.1 |
NM_028763.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014861.mm.1 |
NM_028763.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014861.mm.1 |
TC1500001887.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500014859.mm.1 |
NM_001013362.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014859.mm.1 |
NM_001013362.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014859.mm.1 |
NM_028763.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014859.mm.1 |
NM_028763.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014859.mm.1 |
TC1500001887.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500014856.mm.1 |
NM_028763.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014856.mm.1 |
NM_028763.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014856.mm.1 |
TC1500001887.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500014854.mm.1 |
NM_028763.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014854.mm.1 |
NM_028763.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014854.mm.1 |
TC1500001887.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500014857.mm.1 |
NM_028763.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014857.mm.1 |
NM_028763.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014857.mm.1 |
TC1500001887.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500014860.mm.1 |
NM_001013362.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014860.mm.1 |
NM_001013362.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014860.mm.1 |
NM_028763.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014860.mm.1 |
NM_028763.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014860.mm.1 |
TC1500001887.mm.1 |
MTA TC |