Assay Details
Gene Symbol: Lmbr1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029098.3 | NP_083374.1 | 5 - 6 | 614 | 73 | |
GenBank mRNA | AK003656.1 | - | 5 - 6 | 546 | 73 | |
AK030589.1 | - | 5 - 6 | 580 | 73 | ||
AK049110.1 | - | 5 - 6 | 615 | 73 | ||
AK143891.1 | - | 5 - 6 | 575 | 73 | ||
AK154968.1 | - | 5 - 6 | 748 | 73 | ||
AK170328.1 | - | 5 - 6 | 575 | 73 | ||
AK173121.1 | - | 5 - 6 | 995 | 73 | ||
AY052398.1 | - | 5 - 6 | 457 | 73 | ||
BC023397.1 | - | 5 - 6 | 551 | 5043877 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
