Assay Details
| Gene Symbol: Ascc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029291.1 | NP_083567.1 | 8 - 9 | 878 | 68 | |
| GenBank mRNA | AK048876.1 | - | 8 - 9 | 912 | 68 | |
| AK051387.1 | - | 8 - 9 | 927 | 68 | ||
| AK148105.1 | - | 8 - 9 | 910 | 68 | ||
| BC013537.1 | - | 8 - 9 | 878 | 4240784 | 68 | |
More Information
Set Membership: |
|

Ascc2
Genomic Map