Assay Details
| Gene Symbol: Cyb5r3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029787.2 | NP_084063.1 | 2 - 3 | 228 | 59 | |
| GenBank mRNA | AF332059.1 | - | 2 - 3 | 152 | 59 | |
| AF332060.1 | - | 2 - 3 | 152 | 59 | ||
| AK002640.1 | - | 2 - 3 | 200 | 59 | ||
| AK010858.1 | - | 2 - 3 | 205 | 59 | ||
| AK128917.1 | - | 2 - 3 | 222 | 59 | ||
| AK202011.1 | - | 1 - 2 | 58 | 59 | ||
| AK208840.1 | - | 1 - 2 | 58 | 59 | ||
| BC004760.1 | - | 2 - 3 | 235 | 3154531 | 59 | |
| BC032013.1 | - | 2 - 3 | 210 | 4238275 | 59 | |
| BC043074.1 | - | 2 - 3 | 228 | 6404746 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cyb5r3
Genomic Map