Assay Details
| Gene Symbol: Cyb5r3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029787.2 | NP_084063.1 | 4 - 5 | 408 | 65 | |
| GenBank mRNA | AF332059.1 | - | 4 - 5 | 332 | 65 | |
| AF332060.1 | - | 4 - 5 | 332 | 65 | ||
| AK002640.1 | - | 4 - 5 | 380 | 65 | ||
| AK010858.1 | - | 4 - 5 | 385 | 65 | ||
| AK128917.1 | - | 4 - 5 | 402 | 65 | ||
| AK202011.1 | - | 3 - 4 | 238 | 65 | ||
| AK208840.1 | - | 3 - 4 | 238 | 65 | ||
| BC004760.1 | - | 4 - 5 | 415 | 3154531 | 65 | |
| BC032013.1 | - | 4 - 5 | 390 | 4238275 | 65 | |
| BC043074.1 | - | 4 - 5 | 408 | 6404746 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cyb5r3
Genomic Map